3,5-Dibrom-1-(trimethylsilyl)benzol

Lieferant: CHEMPUR SILANE

Synonyme: 3,5-Dibromophenyltrimethylsilane, (3,5-Dibromophenyl)trimethylsilane

SILS05730.5
SILNSILS05730.5EA 0 EUR
SILNSILS05730.5
3,5-Dibrom-1-(trimethylsilyl)benzol
3,5-Dibrom-1-(trimethylsilyl)benzol
Formel: C₉H₁₂Br₂Si
Molecular Weight: 308.0851 g/mol
MDL: MFCD03844787
CAS-Nummer: 17878-23-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR