1,3-Diethyl-1,1,3,3-tetramethyldisilazan

Lieferant: CHEMPUR SILANE

SILS06525.5
SILNSILS06525.5EA 0 EUR
SILNSILS06525.5
1,3-Diethyl-1,1,3,3-tetramethyldisilazan
1,3-Diethyl-1,1,3,3-tetramethyldisilazane
Formel: C₈H₂₃NSi₂
Molecular Weight: 189,45 g/mol
CAS-Nummer: 17882-94-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR