1,3-Dipropyl-1,1,3,3-tetramethyldisilazan

Lieferant: CHEMPUR SILANE

SILS08275.5
SILNSILS08275.5EA 0 EUR
SILNSILS08275.5
1,3-Dipropyl-1,1,3,3-tetramethyldisilazan
1,3-Dipropyl-1,1,3,3-tetramethyldisilazan
Formel: C₁₀H₂₇NSi₂
Molecular Weight: 217,5 g/mol
Siedepunkt: 205…207 °C (1013 hPa)
Dichte: 0,827 g/cm³ (20 °C)
CAS-Nummer: 14579-90-9
UN: 3267
ADR: 8,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR