1,2-Divinyl-1,1,2,2-tetramethyldisilan

Lieferant: CHEMPUR SILANE

SILS08372.5
SILNSILS08372.5EA 0 EUR
SILNSILS08372.5
1,2-Divinyl-1,1,2,2-tetramethyldisilan
1,2-Divinyl-1,1,2,2-tetramethyldisilan
Formel: C₈H₁₈Si₂
CAS-Nummer: 1450-29-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR