Ethyl(trimethylsilyl)acetat
Lieferant: CHEMPUR SILANE
Synonyme:
(Trimethylsilyl)essigsäureethylester
SILS08900.10
SILS08900.25
SILNSILS08900.10EA
0
EUR
SILNSILS08900.10
SILNSILS08900.25
Ethyl(trimethylsilyl)acetat
Ethyl(trimethylsilyl)acetat
Formel:
C₇H₁₆O₂Si Molecular Weight: 160.29 g/mol |
MDL:
MFCD00009172 CAS-Nummer: 4071-88-9 EINECS: 223-783-2 UN: 3272 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...