(3-Glycidoxypropyl)triethoxysilan

Lieferant: CHEMPUR SILANE

Synonyme: (3-Glycidyloxypropyl)triethoxysilan, Geniosil GF 82, (3-(2,3-Epoxypropoxy)propyl)triethoxysilan

SILS09135.25 SILS09135.100
SILNSILS09135.25EA 0 EUR
SILNSILS09135.25 SILNSILS09135.100
(3-Glycidoxypropyl)triethoxysilan
(3-Glycidoxypropyl)triethoxysilan
Formel: C₁₂H₂₆O₅Si
Molecular Weight: 278.42 g/mol
MDL: MFCD01311705
CAS-Nummer: 2602-34-8
EINECS: 220-011-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR