(Iodmethyl)trimethylsilan
Lieferant: CHEMPUR SILANE
SILS10025.10
SILNSILS10025.10EA
0
EUR
SILNSILS10025.10
(Iodmethyl)trimethylsilan
(Iodomethyl)trimethylsilane
Formel:
C₄H₁₁ISi Siedepunkt: 139 °C (1013 hPa) Dichte: 1,4301 g/cm³ (20 °C) |
MDL:
MFCD00001077 CAS-Nummer: 4206-67-1 EINECS: 224-123-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...