Phenethyltrichlorsilan

Lieferant: CHEMPUR SILANE

SILS13200.10 SILS13200.25 SILS13200.100
SILNSILS13200.10EA 0 EUR
SILNSILS13200.10 SILNSILS13200.25 SILNSILS13200.100
Phenethyltrichlorsilan
Phenethyltrichlorsilan
Formel: C₈H₉Cl₃Si
MDL: MFCD00039290
CAS-Nummer: 940-41-0
EINECS: 213-371-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR