Chlor(dimethyl)phenylsilan
Lieferant: CHEMPUR SILANE
SILS13375.10
SILS13375.25
SILS13375.100
SILNSILS13375.10EA
0
EUR
SILNSILS13375.10
SILNSILS13375.25
SILNSILS13375.100
Chlor(dimethyl)phenylsilan
Chlor(dimethyl)phenylsilan
Formel:
C₈H₁₁ClSi |
MDL:
MFCD00000499 CAS-Nummer: 768-33-2 EINECS: 212-193-0 UN: 2987 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...