Chlor(dimethyl)phenylsilan

Lieferant: CHEMPUR SILANE

SILS13375.10 SILS13375.25 SILS13375.100
SILNSILS13375.10EA 0 EUR
SILNSILS13375.10 SILNSILS13375.25 SILNSILS13375.100
Chlor(dimethyl)phenylsilan
Chlor(dimethyl)phenylsilan
Formel: C₈H₁₁ClSi
MDL: MFCD00000499
CAS-Nummer: 768-33-2
EINECS: 212-193-0
UN: 2987
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR