(Phenylthio)trimethylsilan

Lieferant: CHEMPUR SILANE

SILS13625.5 SILS13625.25
SILNSILS13625.5EA 0 EUR
SILNSILS13625.5 SILNSILS13625.25
(Phenylthio)trimethylsilan
(Phenylthio)trimethylsilane
Formel: C₉H₁₄SSi
MDL: MFCD00008272
CAS-Nummer: 4551-15-9
EINECS: 224-916-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR