Phenyltrimethoxysilan

Lieferant: CHEMPUR SILANE

SILS13750.25 SILS13750.100 SILS13750.250
SILNSILS13750.25EA 0 EUR
SILNSILS13750.25 SILNSILS13750.100 SILNSILS13750.250
Phenyltrimethoxysilan
Phenyltrimethoxysilan
Formel: C₉H₁₄O₃Si
Dichte: 1,064 g/cm³ (20 °C)
MDL: MFCD00025689
CAS-Nummer: 2996-92-1
EINECS: 221-066-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR