Kaliumhexafluorosilicat

Lieferant: CHEMPUR SILANE
Gefahr

Synonyme: Potassium fluorosilicate

SILS13940.500
SILNSILS13940.500EA 0 EUR
SILNSILS13940.500
Kaliumhexafluorosilicat
Potassium hexafluorosilicate
Formel: F₆K₂Si
Molecular Weight: 220,27 g/mol
Dichte: 2,67 g/cm³ (20 °C)
MDL: MFCD00040666
CAS-Nummer: 16871-90-2
EINECS: 240-896-2
UN: 2655
ADR: 6.1,III
Merck Index: 12,07803

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR