Propyltrichlorsilan

Lieferant: CHEMPUR SILANE

Synonyme: Trichlor(propyl)silan, Propylsilyltrichlorid

SILS14075.25 SILS14075.100
SILNSILS14075.25EA 0 EUR
SILNSILS14075.25 SILNSILS14075.100
Propyltrichlorsilan
Propyltrichlorsilan
Formel: C₃H₇Cl₃Si
Molecular Weight: 177.53 g/mol
Siedepunkt: 123.5
Dichte: 1.195 g/cm³ (20 °C)
MDL: MFCD00013603
CAS-Nummer: 141-57-1
EINECS: 205-489-6
UN: 1816
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR