Tetrakis(dimethylsilyl)silan

Lieferant: CHEMPUR SILANE

SILS15680.1
SILNSILS15680.1EA 0 EUR
SILNSILS15680.1
Tetrakis(dimethylsilyl)silan
Tetrakis(dimethylsilyl)silan
Formel: C₈H₂₈Si₅
MDL: MFCD01631315
CAS-Nummer: 2003-85-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR