(Tributylstannyl)trimethylsilan

Lieferant: CHEMPUR SILANE
Gefahr

SILS16775.5
SILNSILS16775.5EA 0 EUR
SILNSILS16775.5
(Tributylstannyl)trimethylsilan
(Tributylstannyl)trimethylsilan
Formel: C₁₅H₃₆SiSn
MDL: MFCD00054903
CAS-Nummer: 17955-46-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR