(Tributylstannylethinyl)trimethylsilan

Lieferant: CHEMPUR SILANE
Gefahr

SILS16776.1
SILNSILS16776.1EA 0 EUR
SILNSILS16776.1
(Tributylstannylethinyl)trimethylsilan
(Tributylstannylethinyl)trimethylsilan
Formel: C₁₆H₃₈SiSn
MDL: MFCD03844806
CAS-Nummer: 81353-38-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR