Azidotrimethylsilan

Lieferant: CHEMPUR SILANE

SILS19050.25
SILNSILS19050.25EA 0 EUR
SILNSILS19050.25
Azidotrimethylsilan
Azidotrimethylsilane
Formel: C₃H₉N₃Si
MDL: MFCD00001986
CAS-Nummer: 4648-54-8
EINECS: 225-078-5
UN: 1992
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR