2-(Trimethylsilyl)benzothiazol

Lieferant: CHEMPUR SILANE

SILS19075.5
SILNSILS19075.5EA 0 EUR
SILNSILS19075.5
2-(Trimethylsilyl)benzothiazol
2-(Trimethylsilyl)benzothiazole
Formel: C₆H₁₁NSSi
MDL: MFCD00068185
CAS-Nummer: 32137-73-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR