4-(Trimethylsilyl)-3-butin-1-ol

Lieferant: CHEMPUR SILANE

Synonyme: 4-Trimethylsilyl-3-butyn-1-ol

SILS19110.5 SILS19110.25
SILNSILS19110.5EA 0 EUR
SILNSILS19110.5 SILNSILS19110.25
4-(Trimethylsilyl)-3-butin-1-ol
4-(Trimethylsilyl)-3-butin-1-ol
Formel: C₇H₁₄OSi
MDL: MFCD00190217
CAS-Nummer: 2117-12-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR