(Trimethylsilylmethyl)triphenylphosphoniumiodid

Lieferant: CHEMPUR SILANE

SILS19850.5
SILNSILS19850.5EA 0 EUR
SILNSILS19850.5
(Trimethylsilylmethyl)triphenylphosphoniumiodid
(Trimethylsilylmethyl)triphenylphosphoniumiodid
Formel: C₂₂H₂₆IPSi
MDL: MFCD00051853
CAS-Nummer: 3739-98-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR