5-(Trimethylsilyl)thiazol

Lieferant: CHEMPUR SILANE

SILS20280.1
SILNSILS20280.1EA 0 EUR
SILNSILS20280.1
5-(Trimethylsilyl)thiazol
5-(Trimethylsilyl)thiazole
Formel: C₆H₁₁NSSi
Molecular Weight: 157,31 g/mol
MDL: MFCD00191506
CAS-Nummer: 79265-36-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR