(Trimethylstannyl)butyldimethylsilan

Lieferant: CHEMPUR SILANE
Gefahr

SILS20410.5 SILS20410.1
SILNSILS20410.5EA 0 EUR
SILNSILS20410.5 SILNSILS20410.1
(Trimethylstannyl)butyldimethylsilan
(Trimethylstannyl)butyldimethylsilan
Formel: C₉H₂₄SiSn
MDL: MFCD03093828
CAS-Nummer: 103731-29-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR