(Trimethylstannyl)triphenylsilan
Lieferant: CHEMPUR SILANE
|
Gefahr
|
SILS20420.1
SILS20420.5
SILNSILS20420.1EA
0
EUR
SILNSILS20420.1
SILNSILS20420.5
(Trimethylstannyl)triphenylsilan
(Trimethylstannyl)triphenylsilan
Formel:
C₂₁H₂₄SiSn |
MDL:
MFCD03093829 CAS-Nummer: 18748-68-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...