Trioctylsilan

Lieferant: CHEMPUR SILANE
Achtung

SILS20500.10 SILS20500.25 SILS20500.100 S20500-25G
SILNSILS20500.10EA 0 EUR
SILNSILS20500.10 SILNSILS20500.25 SILNSILS20500.100 FLUOS20500-25G
Trioctylsilan
Trioctylsilan
Formel: C₂₄H₅₂Si
MDL: MFCD00053812
CAS-Nummer: 18765-09-8
EINECS: 242-559-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR