3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propylmethacrylat

Lieferant: CHEMPUR SILANE

SILS21125.10 SILS21125.25 SILS21125.100
SILNSILS21125.10EA 0 EUR
SILNSILS21125.10 SILNSILS21125.25 SILNSILS21125.100
3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propylmethacrylat
3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propylmethacrylat
Formel: C₁₆H₃₈O₅Si₄
MDL: MFCD00053871
CAS-Nummer: 17096-07-0
EINECS: 241-165-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR