Tris(trimethylsilyl)methan

Lieferant: CHEMPUR SILANE

SILS21300.5
SILNSILS21300.5EA 0 EUR
SILNSILS21300.5
Tris(trimethylsilyl)methan
Tris(trimethylsilyl)methan
Formel: C₁₀H₂₈Si₃
MDL: MFCD00008273
CAS-Nummer: 1068-69-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR