Tris(trimethylsilyl)silan
Lieferant: CHEMPUR SILANE
SILS21375.1
SILS21375.5
SILNSILS21375.1EA
0
EUR
SILNSILS21375.1
SILNSILS21375.5
Tris(trimethylsilyl)silan
Formel:
C₉H₂₈Si₄ Siedepunkt: 82 °C (1013 hPa) Dichte: 0,8 g/cm³ (20 °C) |
MDL:
MFCD00077893 CAS-Nummer: 1873-77-4 Merck Index: 12,09886 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...