Surface probes, clamp-type, for Curvex datalogger
Lieferant: Industrial Physics
CX2049 CX2048
SIMECX2049EA 318.43 EUR
SIMECX2049 SIMECX2048
Surface probes, clamp-type, for Curvex datalogger
Temperaturfühler
Clamp-type probes to measure object temperature (480 °C).
  • Max. object thickness is 25 mm
Bestellen
TECHNISCHE DATEN
Environmentally Preferable