Roughness gauge
Lieferant: Industrial Physics
DC9000
SIMEDC9000EA 5826.17 EUR
SIMEDC9000
Roughness gauge
Beschichtungsprüfgeräte
The (Hull) roughness gauge is a compact system to measure the average Hull roughness (AHR) of sea going vessels, especially for ship operators, ship builders, ship repairers and coating manufacturers.
  • Graphical representations of data and storage in multiple batches
  • Inspection survery reports in Microsoft Excel® using USB-cable and software installed
  • Statistics, time/date and location of series and average hull roughness are calculated and stored
A four direction button operates an intuitive menu on a large display. The neck strap keeps the users' hands free.
The sensor is equipped with three non-slip wheels and a carbide tipped stylus. It is moved over the ship's hull in a horizontal way collecting measurements.
Lieferumfang: Supplied with a neck strap, roughness sensor, sensor cable, calibration plate, thumb drive with software and connection cable for PC.