Roughness gauge
Lieferant: Industrial Physics
LD0015
SIMELD0015EA 3820 EUR
SIMELD0015
Roughness gauge
Beschichtungsprüfgeräte
This roughness meter offers a choice of 39 parameters.
  • Integrated drive unit
  • Internal memory for ten measurements
  • Possible extended storage via a micro SD card
Standards: DIN 3274
Bestellinformation: Includes a data analysis software and cable.