Surface profile comparators
Lieferant: Industrial Physics
LD2053 LD2052 LD2051
SIMELD2053EA 288 EUR
SIMELD2053 SIMELD2052 SIMELD2051
Surface profile comparators
Beschichtungsprüfgeräte
Surface comparators for determining roughness by touch and sight.
  • Five different grades of roughness in each comparator
  • Designed to be used with the illuminated magnifier
  • Available in Imperial units only
Section profiles: 0,5 ; 1,0 ; 2,0 ; 3,0 ; 4,0 mils for the three versions.
Standards: ASTM D4417