Digital resistivity meter for coatings, RE-45
Lieferant: Industrial Physics
LD5950
SIMELD5950EA 3240 EUR
SIMELD5950
Digital resistivity meter for coatings, RE-45
Widerstandstester
Equipment developed to measure the resistivity of paints, varnishes and liquids in a fast and precise way. Indispensable for the correct functioning of electrostatic paint installations.
  • Probe is immersed in the coating and the measured value is displayed on screen
  • Inside Auto scaling allows to measure all ranges of resistivity without changing the reading procedure
  • Readout is displayed in Megaohm (M)
  • Probe (length 230 mm, Ø 42 mm) is easy to clean
  • Helps saving paint and reducing paint waste
Range: 100 Kohm – 20000 Kohm (20 Mohm)
Power supply: 1× 9 V battery
Standards: Meets ASTM D 5682 standards.
Lieferumfang: Supplied in a protective case including verification certificate and manual.