Coating thickness gauge, PosiPen
Lieferant: Industrial Physics
LD6000
SIMELD6000EA 356 EUR
SIMELD6000
Coating thickness gauge, PosiPen
Beschichtungsprüfgeräte
This thickness gauge is designed for the measurement of non magnetic coatings include paint, enamel, plating, hot-dip galvanising on steel. A small magnet can be placed with pin-point accuracy on small parts and on peaks and valleys.
  • Not affected by mechanical shock, acid, oil, water and dust making it rugged
  • Highly wear resistant probe tip for long life and continuous accuracy
  • Two scales: Mils (inch) and microns (metric)
Used for small, hot or hard-to-reach surfaces, spot checks and sprayed coatings. For use in refineries, petrochemical plants, electroplating, chrome, zinc, galvanizing platers and powder coaters.
Lieferumfang: Supplied with a leather pouch, a certificate of calibration (traceable to NIST) and instructions.