Coating thickness gauges, PosiTest DFT
Lieferant: Industrial Physics
LD6011 LD6012
SIMELD6011EA 653 EUR
SIMELD6011 SIMELD6012
Coating thickness gauges, PosiTest DFT
Beschichtungsprüfgeräte
The PosiTest DFT measure coatings on all metals - The DFT Fe measures non-magnetic coatings on steel while the DFT Combo measures both non magnetic coatings on steel and non conductive coatings on aluminum and brass.
  • Zero feature for rough or curved surfaces with one point calibration adjustment for known thicknesses
  • Displays the moving average for up to the last ten measurements
  • Handy reset feature when no zero reference is available
  • FLIP display enables right side up viewing with audible and visible measurement indication
  • V-groove in probe for positioning on cylindrical parts with built-in wrist strap for added convenience and safety
  • Mils/microns switchable with basic instructions on the back of each gauges
  • Automatically recognises the substrate and takes a measurement (DFT Combo)
Standards: Traceability to NIST.
Lieferumfang: Supplied with built-in probe, wrist strap, plastic shims, hard shell storage case, AAA alkaline battery, instructions, protective lens shield, long form certificate of calibration.