Thickness gauges
Lieferant: Industrial Physics
LD7016
SIMELD7016EA 649.68 EUR
SIMELD7016
Thickness gauges
Beschichtungsprüfgeräte
These thickness gauges are designed to measure the thickness of metallic and non-metallic materials (aluminium, titanium, plastics, ceramics, glass, plastics). It can monitor pipes and vessels for loss of thickness due to corrosion or erosion.
  • Large screen, battery indicator and a coupling indicator
  • Automatic power shut down, simple calibration procedure
  • Not suitable for cast iron due to its big crystalloid composition
'Basic' model has ten preset sound velocities.
'Pro' version allows for the storage of data (10 storage units) and performs a sound velocity measurement thanks to a known material thickness.
Lieferumfang: Supplied with ultrasonic gel (60 ml), probe (10 mm, 5,0 MHz), calibration block and manual. 'Pro' version has an extra probe (10 mm, 2,5 MHz).