High voltage holiday detectors
Lieferant: Industrial Physics
LD8504 LD8503
SIMELD8504EA 3177.27 EUR
SIMELD8504 SIMELD8503
High voltage holiday detectors
Beschichtungsprüfgeräte
This apparatus is designed to detect of coating porosity in dielectric (insulation type) coatings on conductive substrates, including concrete.
  • Digital display of applied voltage with integral battery condition indicator
  • Regulated DC voltage with adjustable ranges
  • Momentary on switch allows auto shut off with clip on battery pack
  • Sensitivity control with power supply
  • Constant test current and overcharge protection
  • Earphones for noisy environments and optional On/Off switch
  • Audible and visual alarm
Detector provides accurate detection of pinholes,More Product Information