Wood moisture meters, MaxiLigno
Lieferant: Industrial Physics
LI0010
SIMELI0010EA 1100 EUR
SIMELI0010
Wood moisture meters, MaxiLigno
Analysatoren Feuchte-Analysatoren
The MaxiLigno are equipped with the latest technology for precision and easy handling.
  • Different languages: English, French, German, Swedish
  • Large illuminated display shows the wood species and temperature
  • Block battery (9 V) sufficient for 3000 measurements
They can offer analyses for 28 woods and all 15 density groups for more than 240 wood species.
Lieferumfang: Supplied with a cable and a case.