Non-destructive moisture meter, Encounter Plus Tramex®
Lieferant: Industrial Physics
LI9010
SIMELI9010EA 624.02 EUR
SIMELI9010
Non-destructive moisture meter, Encounter Plus Tramex®
Analysatoren Feuchte-Analysatoren
Encounter Plus is a non-destructive moisture meter designed fo the inspection in numerous industries.
  • High moisture audio warning tone
  • Precise and versatile evaluation and measurements
  • Latest digital technology with analogue read out and ease of use
Designed for detecting leaks in roofs or dry walls, wood and construction materials.
Lieferumfang: Supplied in a case.