Wood and plastic moisture tester, Skipper Plus
Lieferant: Industrial Physics
LI9012
SIMELI9012EA 593.7 EUR
SIMELI9012
Wood and plastic moisture tester, Skipper Plus
Analysatoren Feuchte-Analysatoren
Skipper Plus is a non-destructive moisture detector designed for locating damp in wood or laminated plastics such as polyester.
  • Short rubber electrodes, which do not damage the surface
  • Three moisture ranges for moisture in both fresh and salt water
  • Developed for osmosis-control and measuring moisture before painting yachts
W×D×H: 150×80×30 mm