Digital thermo-hygrometer
Lieferant: Industrial Physics
RV2100
SIMERV2100EA 234.45 EUR
SIMERV2100
Digital thermo-hygrometer
Hygrometer Thermohygrometer, digital
Simple hand-held digital thermo/hygrometer.
  • Temperature and air humidity are displayed simultaneously
  • Operates 10000 hours on three AAA batteries and switches itself off after 10 min
  • Equipped with Min./Max. memory, C°/F° selection, HOLD function, internal probe and dewpoint indication
Lieferumfang: Supplied in a soft pouch with manual and batteries.