Surface roughness gauge
Lieferant: Industrial Physics
SP1562
SIMESP1562EA 443.23 EUR
SIMESP1562
Surface roughness gauge
Beschichtungsprüfgeräte
Surface profile roughness gauge for ultrafast measurement of the peaks and valleys of a surface.
The shape of the base is suitable to measure the height of welds.
Standards: ASTM D4417
Lieferumfang: Supplied with a reference glass plate, an Allen key and an extra battery in a leather pouch.