Wet film thickness gauges
Lieferant: Industrial Physics
SP4010 SP4020 SP4000 SP4060
SIMESP4010EA 62.99 EUR
SIMESP4010 SIMESP4020 SIMESP4000 SIMESP4060
Wet film thickness gauges
Beschichtungsprüfgeräte
Hexagonal/octagonal precision measuring comb made of heavy stainless steel - The high-grade stainless steel will not be affected by acid or base elements.
  • Available in microns and mils scale
  • For decorative paints and primers (20 to 370 µm)
  • For protective coatings and coating with high solids content (25 to 2000 µm)
  • For extremely thick films such as floor coating, fillers, fire proofing, plasters and adhesives (50 to 10000 µm)
  • For protective coatings and coating with high solids content (1 to 80 mils)