Wedge for measuring reverse-side tolerance
Lieferant: Industrial Physics
SP6000
SIMESP6000EA 69.98 EUR
SIMESP6000
Wedge for measuring reverse-side tolerance
Mess- und Nivelliergeräte
Anodised aluminum wedge for quick measurement of reverse-side tolerance of windows and doors.
  • Tolerance is given in millimetres
  • Also equipped with a measuring scale for optically determining the thickness or glass