Magnetic thermometer
Lieferant: Industrial Physics
TM0015
SIMETM0015EA 83.98 EUR
SIMETM0015
Magnetic thermometer
Thermometer Analoge Thermometer
Magnetic thermometer for measuring the surface temperature of steel objects. The thermometer can easily be placed on ferrous substrates with the strong magnet on the back.
  • Surface temperature can be read out in both °C and °F
  • Can be used in vacuum
  • No plastic parts on the outside of the thermometer
Lieferumfang: Supplied in protective leather pouch with belt-clip.