Micrometer Film Applicators
Lieferant: Industrial Physics
VF1820 VF1821 VF1822 VF1823 VF1825 VF1824
SIMEVF1820EA 1220 EUR
SIMEVF1820 SIMEVF1821 SIMEVF1822 SIMEVF1823 SIMEVF1825 SIMEVF1824
Micrometer Film Applicators
Filmaufziehgeräte Verschlussfolienapplikator
Adjustable Micrometer Film Applicator. The applicator blade can be adjusted in 10 μm increments by means of 2 μm heads, over a gap size from 0 to 8 mm.
  • Stainless steel knife blade with precision ground edge
  • Wet film thickness adjustable to 8 mm maximum
  • Micrometer head accuracy is ±10 µm
  • Ergonomic design facilitates fast accurate draw downs
  • Anodised aluminium material, through hardened medical grade stainless steel
  • Black anodised aluminium frame provides a large paint well
The applicator offers the user flexibility to adjust the optimum gap size forMore Product Information