Permeability cups
Lieferant: Industrial Physics
VF2200 VF2201
SIMEVF2200EA 221.62 EUR
SIMEVF2200 SIMEVF2201
Permeability cups
Probenbehälter Probengefäße für Analyser
Permeability cups are designed to determine the water vapour transmission of paints, varnishes, coatings and related products. Permeability cup is suitable for testing both self-supporting coating and non-self-supporting coatings.
  • Anodised aluminium for corrosion resistance and colouring
  • Seal ring to prevent turning when closing the cover
  • Any deformation to the cup may cause deviation in the result
The wet cup method is the reference for determining water vapour transmission.
Standards: ISO 7783 (supersedes NF T30-018), ASTM D1653, ASTM E95.