Colour reference, RAL books for hotel
Lieferant: Industrial Physics
VF6643
SIMEVF6643EA
48.3
EUR
SIMEVF6643
Colour reference, RAL books for hotel
Standards
Farbreferenzen und Spektroskopie-Standards
RAL book colours for hotels.
- Coded in the RAL DESIGN system
- Based on a scientific study, includes German/English languages
- For architects, designers, investors, building owners, hotel managers and advertisers in the hotel industry
- With practical and relevant design examples
- Includes a template for colour combinations