Scratching tools, Machu
Lieferant: Industrial Physics
VF8605 VF8600
SIMEVF8605EA 530.71 EUR
SIMEVF8605 SIMEVF8600
Scratching tools, Machu
Schneidewerkzeuge
These scratching tools can perform a Machu corrosion test. Each tool is provided with a 1 mm width cutter to cut the coating down to the metal.
  • Ergonomic shaped grip
  • Easy to replace the cutter
  • Mandatory test in Qualicoat and QIB accredited laboratories
CC2000 has a self positioning knife bracket to expose the substrate with a perpendicular cut through the coating. CC3000 has an adjustable cutting depth and two ball bearings to guide the cut and this guarantees reproducible results.
Standards: ISO 17872. DIN 53167, 50021, ISO 4628-8, 9227.