Magnetic hotplate stirrer, digital
Lieferant: Industrial Physics
VF8800
SIMEVF8800EA 992.61 EUR
SIMEVF8800
Magnetic hotplate stirrer, digital
Rührer Magnetrührer mit Heizplatte
This ceramic coated hotplate stirrer has high chemical and acid resistance.
  • Illuminated digital display shows values
  • Values can be stored in memory
  • Jog-Shuttle switch (turn and push) eases heat setting (up to 380 °C), stirring speed (80 to 1500 min⁻¹) and time
  • Protected against overheating and overload
The end of a run or probe failure is indicated with an acoustic signal.
Lieferumfang: Supplied with external temperature probe, support rod and clamp.